Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cesium, Ultrapure, Spectrum™

Click to view available options
Quantity:
1 g
5 g
7440-46-2
7440-46-2
Quantity 5 g
CAS Max % 1
Grade Ultrapure

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.