Learn More
HyClone™ CET Human Adipose-Derived Mesenchymal Stem Cells
Manufacturer:  HyClone™ SV30102.01
 View more versions of this product
Catalog No. SV3010201
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
HyClone™ CET Human Adipose-Derived Mesenchymal Stem Cells are isolated and expanded from human lipoaspirate using enzymatic treatments.
Specifications
Specifications
CET Human Adipose-Derived Mesenchymal Stem Cells = 100,000
Store at -196°C in liquid nitrogen.
Human Adipose-Derived Mesenchymal Stem Cells
2mL