Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HyClone™ CET Human Adipose-Derived Mesenchymal Stem Cells

Specifications

Quantity 2mL
Storage Store at -196°C in liquid nitrogen.
Cell Type Human Adipose-Derived Mesenchymal Stem Cells
View More Specs

Products 1
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
SV3010201 View Documents HyClone™
SV30102.01
CET Human Adipose-Derived Mesenchymal Stem Cells = 100,000 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

HyClone™ CET Human Adipose-Derived Mesenchymal Stem Cells are isolated and expanded from human lipoaspirate using enzymatic treatments.

Specifications

Specifications

2mL
Store at -196°C in liquid nitrogen.
Human Adipose-Derived Mesenchymal Stem Cells
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit