Learn More
HyClone™ CET Human Bone Marrow Mesenchymal Stem Cells
Manufacturer:  HyClone™ SV30110.01
 View more versions of this product
Catalog No. SV3011001
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
HyClone™ CET Human Bone Marrow Mesenchymal Stem Cells are isolated from human red bone marrow collected during a bone marrow aspiration procedure.
Specifications
Specifications
100,000 cells/mL
Store at -196°C in liquid nitrogen.
Human Bone Marrow Mesenchymal Stem Cells
2mL