Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cetirizine dihydrochloride, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
50mg
This item is not returnable. View return policy
83881-52-1
C21H27Cl3N2O3
461.808
PGLIUCLTXOYQMV-UHFFFAOYSA-N
cetirizine dihydrochloride,zyrtec,reactine,cetirizine hydrochloride,2-2-4-4-chlorophenyl phenyl methyl piperazin-1-yl ethoxy acetic acid dihydrochloride,alerlisin,alercet,alergex,alertisin,cetriler
55182
2-[2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]acetic acid;dihydrochloride
C1CN(CCN1CCOCC(=O)O)C(C2=CC=CC=C2)C3=CC=C(C=C3)Cl.Cl.Cl
This item is not returnable. View return policy
83881-52-1
C21H27Cl3N2O3
461.808
PGLIUCLTXOYQMV-UHFFFAOYSA-N
cetirizine dihydrochloride,zyrtec,reactine,cetirizine hydrochloride,2-2-4-4-chlorophenyl phenyl methyl piperazin-1-yl ethoxy acetic acid dihydrochloride,alerlisin,alercet,alergex,alertisin,cetriler
55182
2-[2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]acetic acid;dihydrochloride
C1CN(CCN1CCOCC(=O)O)C(C2=CC=CC=C2)C3=CC=C(C=C3)Cl.Cl.Cl

Selective H1 antagonist

Histamine H 1 receptor antagonist that displays selectivity over other receptors at concentrations up to 10 μM. A non-sedating antihistamine that inhibits histamine release and eosinophil chemotaxis during secondary phase allergic response. Inhibits activation of eosinophils, neutrophils and monocytes in vivo.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 50mg
Formula Weight 461.81
Percent Purity >99%
Chemical Name or Material Cetirizine dihydrochloride

Recommended Storage : Desiccate at Room Temperature