Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cetostearyl Alcohol, NF, 90-102%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
8005-44-5
C34H72O2
512.95
UBHWBODXJBSFLH-UHFFFAOYSA-N
hexadecan-1-ol; octadecan-1-ol
CCCCCCCCCCCCCCCCO.CCCCCCCCCCCCCCCCCCO
8005-44-5
C34H72O2
512.95
UBHWBODXJBSFLH-UHFFFAOYSA-N
hexadecan-1-ol; octadecan-1-ol
CCCCCCCCCCCCCCCCO.CCCCCCCCCCCCCCCCCCO

C1888, 8005-44-5

Spectrum™ Cetostearyl Alcohol, NF is used as a stabilizer and opacifying agent. The NF grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Quantity 2.5 kg
Packaging Poly Pail
CAS Max % 100%
Percent Purity 90-102%