Learn More
Cetuximab, Rabbit anti-Many, Alexa Fluor 350, Clone: 2259C, R&D Systems™
Rabbit Monoclonal Antibody
Manufacturer:  R&D Systems FAB9626U100UG
Catalog No. FAB9626U100
Add to cart
This item is not returnable. View return policy
Description
Description
Cetuximab Monoclonal antibody specifically detects Cetuximab in Multi-Species Samples. It is validated for Flow Cytometry.
Specifications
Specifications
Cetuximab
2259C
Flow Cytometry 0.25-1 ug/10^6 cells
Purified
Full length Cetuximab,
Protein A or G purified from cell culture supernatant
RUO
Primary
Detects Cetuximab in Western blots and direct ELISAs. No cross-reactivity with Adalimumab, Rituximab, Trastuzumab, Brentuximab, Gemtuzumab, Ibritumomab, Tositumomab, Solanezumab, Teprotumumab, and Aducanumab is observed.
Flow Cytometry
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Rabbit
IgG
100 ug
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Many