Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cetylpyridinium Chloride, Monohydrate, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
50 kg
Packaging:
Glass Bottle
Plastic Bottle
Steel Drum
6004-24-6
C21H40ClNO
358.01
NFCRBQADEGXVDL-UHFFFAOYSA-M
1-hexadecyl-1,4-dihydropyridin-4-ylium hydrate chloride
O.[Cl-].CCCCCCCCCCCCCCCCN1C=C[CH+]C=C1
6004-24-6
C21H40ClNO
358.01
NFCRBQADEGXVDL-UHFFFAOYSA-M
1-hexadecyl-1,4-dihydropyridin-4-ylium hydrate chloride
O.[Cl-].CCCCCCCCCCCCCCCCN1C=C[CH+]C=C1

CE121, 6004-24-6, C21H38ClN•H2O

Spectrum™ Cetylpyridinium Chloride, Monohydrate, USP is used as an oral antiseptic. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Ignition Residue 0.2%
Quantity 500 g
Packaging Glass Bottle
CAS Max % 100%
UN Number UN2811
Percent Purity 98-102%