Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cetyltrimethylammonium Bromide, 98-101%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
57-09-0
C19H42BrN
364.46
LZZYPRNAOMGNLH-UHFFFAOYSA-M
hexadecyltrimethylazanium bromide
[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C
57-09-0
C19H42BrN
364.46
LZZYPRNAOMGNLH-UHFFFAOYSA-M
hexadecyltrimethylazanium bromide
[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C

CE122, 57-09-0, C19H42BrN

Spectrum™ Cetyltrimethylammonium Bromide is also referred to as CTAB and is a cationic surfactant that is used as a buffer solution during the extraction of DNA, in hair conditioning products, as well as, in the synthesis of gold nanoparticles.
Packaging Amber Glass Bottle
Quantity 500 g
CAS Max % 100%
Percent Purity 98 to 101%