Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CFAP126 Rabbit anti-Human, HRP, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298012H
Description
CFAP126 Polyclonal specifically detects CFAP126 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CFAP126 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
chromosome 1 open reading frame 192 | |
Produced in rabbits immunized with E. coli-derived Human CFAP126 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HRP | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
257177 | |
Store at 4°C in the dark. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title