Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CFDP1 Mouse anti-Human, Clone: GT11811, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527809

Catalog No. PIMA527809


Add to cart

Description

Description

Positive Control: 293T, U87-MG, SK-N-SH, IMR32, SK-N-AS Predicted Reactivity: Dog (89%), Rhesus Monkey (98%), Bovine (80%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

CFDP1 may play a role during embryogenesis.
Specifications

Specifications

CFDP1
Monoclonal
2.34 mg/ml
PBS with no preservative
Q9UEE9
CP27, SWC5, CENP-29, Yeti, BCNT, p97, craniofacial development protein 1, BUCENTAUR
Mouse
IgG2a
100 μl
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
10428
Western Blot
GT11811
Unconjugated
CFDP1
Liquid
CFDP1
Recombit protein encompassing a sequence within the center region of human CFDP1. The exact sequence is proprietary.
Protein G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit