Learn More
Azzota Corp CFI-400945 DFS Item
Manufacturer:  Azzota Corp C1105
CFI-400945 is an orally available PLK4 (polo-like kinase 4) inhibitor with potential antineoplastic activity. CFI-400945 selectively inhibits PLK4, which results in the disruption of mitosis and the induction of apoptosis. Please request if need more quantities
Catalog No. 50-161-3088