Learn More
CFI Mouse anti-Human, Clone: OTI11G3, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF806063
Catalog No. 50-166-7678
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Factor I, a serine protease, is a regulatory component of both the alternative and the classical complement pathway. Factor I is synthesized as a single chain precursor of 88 kDa, processed to yield the acitve form, composed of two disulfide-linked chains, the a-chain (50 kDa) and the beta-chain (38 kDa). Plasma concentration is 35 ug/ml.
Specifications
Specifications
CFI
Monoclonal
Unconjugated
CFI
Lyophilized
CFI
Human recombit protein fragment corresponding to amino acids 340-583 of human CFI produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI11G3
PBS with 8% trehalose and no preservative; pH 7.3
P05156
AHUS3, ARMD13, C3BINA, C3b-INA, FI, IF, KAF
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
3426