Learn More
Aviva Systems Biology Corp CFLAR Antibody - N-terminal region (AVARP00022_T100) | 100uL
Supplier:  Aviva Systems Biology Corp AVARP00022_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CFLAR. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CFLAR. Alternate names: CASH, FLIP, MRIT, CLARP, FLAME, Casper, FLAME1, c-FLIP, FLAME-1, I-FLICE, c-FLIPL, c-FLIPR, c-FLIPS, CASP8AP1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rat
Catalog No. 50-014-62454