Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CFTR Mouse anti-Human, Alexa Fluor™ 350, Clone: CFTR/1644, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP254318AF350
Description
CFTR Monoclonal antibody specifically detects CFTR in Human samples. It is validated for ELISA,Immunocytochemistry,Immunofluorescence.Specifications
CFTR | |
CFTR/1644 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
ABC35, ABCC7CF, ATP-binding cassette sub-family C member 7, ATP-binding cassette transporter sub-family C member 7, cAMP-dependent chloride channel, CFTR/MRP, Channel conductance-controlling ATPase, cystic fibrosis transmembrane conductance regulator, cystic fibrosis transmembrane conductance regulator (ATP-binding cassettesub-family C, member 7), cystic fibrosis transmembrane conductance regulator, ATP-binding cassette(sub-family C, member 7), dJ760C5.1, EC 3.6.3, MRP7EC 3.6.3.49, TNR-CFTR | |
Mouse | |
IgG1 | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
ELISA, Immunocytochemistry, Immunofluorescence | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
CFTR | |
Recombinant human CFTR fragment (aa258-385) (exact sequence is proprietary) | |
0.1 mL | |
Primary | |
1080 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title