Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CFTR Mouse anti-Human, Alexa Fluor™ 750, Clone: CFTR/1644, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254318AF750

Catalog No. NP24318A70


Add to cart

Description

Description

CFTR Monoclonal antibody specifically detects CFTR in Human samples. It is validated for ELISA,Immunocytochemistry,Immunofluorescence.
Specifications

Specifications

CFTR
CFTR/1644
ELISA, Immunocytochemistry/Immunofluorescence
ABC35, ABCC7CF, ATP-binding cassette sub-family C member 7, ATP-binding cassette transporter sub-family C member 7, cAMP-dependent chloride channel, CFTR/MRP, Channel conductance-controlling ATPase, cystic fibrosis transmembrane conductance regulator, cystic fibrosis transmembrane conductance regulator (ATP-binding cassettesub-family C, member 7), cystic fibrosis transmembrane conductance regulator, ATP-binding cassette(sub-family C, member 7), dJ760C5.1, EC 3.6.3, MRP7EC 3.6.3.49, TNR-CFTR
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Immunocytochemistry, Immunofluorescence
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
CFTR
Recombinant human CFTR fragment (aa258-385) (exact sequence is proprietary)
0.1 mL
Primary
1080
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit