Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CFTR Mouse anti-Human, Mouse, Clone: CFTR/1342, Abnova™

Mouse Monoclonal Antibody

Supplier:  Abnova Corporation MAB14728

Catalog No. 89-034-811


Add to Cart

Description

Description

Mouse monoclonal antibody raised against full length recombinant human CFTR.

  • High specificity
  • High reactivity
Specifications

Specifications

CFTR
Monoclonal
Unconjugated
Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
P13569
CFTR
Recombinant protein corresponding to full length human CFTR.
100 μg
Primary
Human, Mouse
Liquid
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
CFTR/1342
Mouse monoclonal antibody raised against full length recombinant human CFTR.
In 10 mM PBS.
ABC35/ABCC7/CF/CFTR/MRP/MRP7/TNR-CFTR/dJ760C5.1
Mouse
Protein A/G purification
RUO
1080
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
IgG2a
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)