Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CFTR Mouse anti-Human, Mouse, Clone: rCFTR/1342, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30749510UG
Description
CFTR Monoclonal specifically detects CFTR in Human, Mouse samples. It is validated for Immunohistochemistry-Paraffin.Specifications
CFTR | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
ABC35, ABCC7CF, ATP-binding cassette sub-family C member 7, ATP-binding cassette transporter sub-family C member 7, cAMP-dependent chloride channel, CFTR/MRP, Channel conductance-controlling ATPase, cystic fibrosis transmembrane conductance regulator, cystic fibrosis transmembrane conductance regulator (ATP-binding cassettesub-family C, member 7), cystic fibrosis transmembrane conductance regulator, ATP-binding cassette(sub-family C, member 7), dJ760C5.1, EC 3.6.3, MRP7EC 3.6.3.49, TNR-CFTR | |
Recombinant full-length human CFTR protein. | |
100 μg | |
ABC Transporters, Cancer, Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones | |
1080 | |
Store at 4°C. | |
IgG1 κ |
Immunohistochemistry (Paraffin) | |
rCFTR/1342 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title