Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CFTR Antibody (CFTR/1775R), Novus Biologicals™
Rabbit Monoclonal Antibody
$313.00 - $566.50
Specifications
Antigen | CFTR |
---|---|
Clone | CFTR/1775R |
Concentration | 0.2 mg/mL |
Dilution | Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB25337920
|
Novus Biologicals
NBP25337920UG |
20 μg |
Each for $313.00
|
|
|||||
NB253379100
|
Novus Biologicals
NBP253379100UG |
100 μg |
Each for $566.50
|
|
|||||
NB253379200
|
Novus Biologicals
NBP253379200UG |
200 μg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
CFTR Monoclonal specifically detects CFTR in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CFTR | |
0.2 mg/mL | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
ABC Transporters, Cancer, Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones | |
10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
1080 | |
Recombinant human CFTR fragment (aa258-385) (exact sequence is proprietary) | |
Primary | |
Store at 4C. |
CFTR/1775R | |
Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
ABC35, ABCC7CF, ATP-binding cassette sub-family C member 7, ATP-binding cassette transporter sub-family C member 7, cAMP-dependent chloride channel, CFTR/MRP, Channel conductance-controlling ATPase, cystic fibrosis transmembrane conductance regulator, cystic fibrosis transmembrane conductance regulator (ATP-binding cassettesub-family C, member 7), cystic fibrosis transmembrane conductance regulator, ATP-binding cassette(sub-family C, member 7), dJ760C5.1, EC 3.6.3, MRP7EC 3.6.3.49, TNR-CFTR | |
CFTR | |
IgG κ | |
Protein A purified | |
167 kDa |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title