Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CFTR Rabbit anti-Human, Mouse, Unconjugated, Clone: CFTR/2290R, Abnova™

Rabbit Monoclonal Antibody.

Manufacturer:  Abnova Corporation RAB00536

Catalog No. 89-339-492


Add to cart

Description

Description

CFTR Monoclonal antibody specifically detects CFTR in Human, Mouse samples. It is validated for IHC-P.
Specifications

Specifications

CFTR
CFTR/2290R
cystic fibrosis transmembrane conductance regulator (ATP-binding cassette sub-family C, member 7)
Liquid
Rabbit
IgG
Store at -20 to -80°C
Monoclonal
Human, Mouse
Immunohistochemistry (Paraffin)
Unconjugated
CFTR
ABC35, ABCC7, CF, CFTR/MRP, MRP7, TNR-CFTR, dJ760C5.1
Recombinant protein corresponding to full-length human CFTR
100 μg
Primary
1080
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit