Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CFUrine Kit™ Available on GSA/VA Contract for Federal Government customers only.

CFUrine quality control kits are used for determination of urine colony counts for dipslides, tube screening methods and calibrated loops.

$116.10 - $125.58

Specifications

Target Organism Class Organism Sets
Format Vials
View More Specs

Products 2
Catalog Number Mfr. No. ATCC® Description Price Quantity & Availability  
Catalog Number Mfr. No. ATCC® Description Price Quantity & Availability  
R4702000
View Documents
Thermo Scientific™
R4702000
12453TM* CFUrine Kit™ Proteus mirabilis Pack for $125.58
Add to cart
 
R4703000 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R4703000
25923TM* CFUrine Kit™ Staphylococcus aureus subsp. aureus Pack for $116.10
Add to cart
 
Description

Description

CFUrine quality control kits are used for determination of urine colony counts for dipslides, tube screening methods and calibrated loops.

  • Film of preserved microorganisms on the inside of the cap on plastic vial
  • Ready for rehydration

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Organism Sets
Vials
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit