Learn More
CGA Mouse anti-Human, Clone: ME.111, Abnova™
Mouse monoclonal antibody raised against human CGA.
Manufacturer:  Abnova Corporation MAB12729
Catalog No. 89-933-965
Add to cart
Description
Description
The four human glycoprotein hormones chorionic gonadotropin (CG), luteinizing hormone (LH), follicle stimulating hormone (FSH), and thyroid stimulating hormone (TSH) are dimers consisting of alpha and beta subunits that are associated noncovalently. The alpha subunits of these hormones are identical, however, their beta chains are unique and confer biological specificity. The protein encoded by this gene is the alpha subunit and belongs to the glycoprotein hormones alpha chain family. [provided by RefSeq]
Specifications
Specifications
CGA
ME.111
Mouse monoclonal antibody raised against human CGA.
In PBS, pH 7.2 (40% glycerol).
CG-ALPHA/FSHA/GPHA1/GPHa/HCG/LHA/TSHA
Mouse
IgG1, kappa
100 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot The optimal working dilution should be determined by the end user.
Liquid
CGA
Human CGA
Protein A purification
RUO
Primary
1081