Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cGAS Mouse anti-Human, Clone: CL3605, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP261624

Catalog No. NBP261624


Add to cart

Description

Description

cGAS Monoclonal antibody specifically detects cGAS in Human samples. It is validated for Western Blot.
Specifications

Specifications

cGAS
CL3605
Western Blot 1 ug/ml
Purified
MB21D1
This antibody was developed against a recombinant protein corresponding to amino acids: RKQLRLKPFYLVPKHAKEGNGFQEETWRLSFSHIEKEILNNHGKSKTCCENKEEKCCRKDCLKLMKYLLEQLKERF
Protein A purified
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Western Blot
Unconjugated
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
C6orf150, c-GAS, cyclic GMP-AMP synthase, h-cGAS, Mab-21 domain containing 1
Mouse
IgG1
100 ul
Primary
115004
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit