Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CGB Rabbit anti-Human, Unconjugated, Clone: HCGb/1985R, Abnova
Rabbit Monoclonal Antibody.
Manufacturer: Abnova Corporation RAB00392
Description
CGB Monoclonal antibody specifically detects CGB in Human samples. It is validated for IHC-P.Specifications
CGB | |
HCGb/1985R | |
chorionic gonadotropin, beta polypeptide | |
Liquid | |
Rabbit | |
IgG | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. | |
Monoclonal | |
Human |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
CGB | |
CGB3, hCGB | |
Original antibody is raised against recombinant protein corresponding to full length human CGB | |
100 μg | |
Primary | |
1082 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title