Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cGKII Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550931

Catalog No. PA550931


Add to cart

Description

Description

cGKII Polyclonal Antibody detects endogenous levels of cGKII protein.

This gene encodes a protein that belongs to the serine/threonine protein kinase family of proteins. The encoded protein plays a role in the regulation of fluid balance in the intestine. A similar protein in mouse is thought to regulate differentiation and proliferation of cells in the colon. Alternate splicing results in multiple transcript variants. [provided by RefSeq, Sep 2013]
Specifications

Specifications

cGKII
Polyclonal
Unconjugated
PRKG2
Liquid
PRKG2
Synthesized peptide derived from human cGKII around the non-phosphorylation site of S126.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
ELISA, Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Q13237, Q61410, Q64595
PRKG2, PRKGR2, cGMP-dependent protein kinase 2, cGK 2, cGK2, cGMP-dependent protein kinase II, cGKII
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
19092, 25523, 5593
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit