Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cGKII Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549822

Catalog No. PA549822


Add to cart

Description

Description

The antibody detects endogenous levels of total cGKII protein.

cGKII is thought to play a key role in a diverse set of physiological pathway. cGKII may mediate intestinal secretion of water and electrolytes induced by the E. coli toxin STa and the intestinal peptide guanylin. Edentification of the pathway that mediates intestinal fluid secretion by E. coli STa has potential medical implications because STa causes traveler ft.s diarrhea and about 50% of infant mortality in developing countries. Transfection experiments in human cells disclose that cGKII phosphorylates SOX9 and attenuates SOX9 function by inhibiting its nuclear entry. Impaired differentiation of cultured KMI chondrocytes can be restored by silencing Sox9 by RNA interference. cGKII is postulated to be a molecular switch that couples the cessation of proliferation and the start of hypertrophic chondrocyte differentiation through attenuating SOX9 function.
Specifications

Specifications

cGKII
Polyclonal
Unconjugated
PRKG2
Liquid
PRKG2
Synthesized peptide derived from internal of human CGK 2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q13237, Q61410, Q64595
cGMP-dependent protein kinase 2, EC 2.7.11.12, CGK 2, Cgkii, Type II cGMP-dependent protein kinase
Rabbit
IgG
100 μL
-20°C
Primary
19092, 25523, 5593
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit