Learn More
Tocris Bioscience™ cGMP dependent kinase inhibitor peptide
Inhibitor of protein kinases G and A
Supplier:  Tocris Bioscience™ 1883/1
Catalog No. 18-831-R
Add to cart
This item is not returnable. View return policy
Description
Description
cGMP Dependent Kinase Inhibitor Peptide is a competitive inhibitor of cGMP-dependent protein kinase (PKG); analog of a substrate peptide corresponding to a phosphorylation site of histone H2B. Competes with synthetic substrates (Ki = 86 mM) but does not inhibit phosphorylation of intact histones by PKG. Inhibits phosphorylation of intact histones by PKA.
Specifications
Specifications
cGMP dependent kinase inhibitor peptide
cGMP Dependent Kinase Inhibitor Peptide
Arg-Lys-Arg-Ala-Arg-Lys-Glu
943.12
1 mg