Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CGN Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555673
Description
Immunogen sequence: DSKQALQQLQ AQLEDYKEKA RREVADAQRQ AKDWASEAEK TSGGLSRLQD EIQRLRQALQ ASQAERDTAR LDKELLAQRL QGLEQEAENK KRSQDDRARQ LKGLEE Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
Probably plays a role in the formation and regulation of the tight junction (TJ) paracellular permeability barrier. [UniProt].
Specifications
CGN | |
Polyclonal | |
Unconjugated | |
CGN | |
6330408J11Rik; CGN; Cingulin; KIAA1319 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
310655, 57530 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P2M7 | |
CGN | |
Recombinant protein corresponding to Human CGN. Recombinant protein control fragment (Product #RP-94283). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction