Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CGP 3466B maleate, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
CGP 3466B maleate is an orally active glyceraldehyde-3-phosphate dehydrogenase (GAPDH) inhibitor. Exhibits antiapoptotic activity via inhibition of GAPDH nuclear translocation and mitochondrial complex 1-dependent H2O2 release. CGP 3466B inhibits colon cancer proliferation in vitro and displays neuroprotective activity in vivo.
Specifications
Quantity | 10 mg |
Formula Weight | 391.42 |
Percent Purity | >99% |
Chemical Name or Material | CGP 3466B maleate |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction