Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CGP 52411, Tocris Bioscience™

Catalog Number 336010
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
145915-58-8
C20H15N3O2
329.36
MFCD00236442
AAALVYBICLMAMA-UHFFFAOYSA-N
daph, 4,5-dianilinophthalimide, 5,6-bis phenylamino-1h-isoindole-1,3 2h-dione, 5,6-dianilino-1h-isoindole-1,3 2h-dione, unii-z13d008fz2, d03qtp, daph-1, 4,5-bis anilino phthalimide, 4,5-bis-anilino phthalimide, 1h-isoindole-1,3 2h-dione, 5,6-bis phenylamino
1697
CHEBI:53110
5,6-bis(phenylamino)-2,3-dihydro-1H-isoindole-1,3-dione
O=C1NC(=O)C2=CC(NC3=CC=CC=C3)=C(NC3=CC=CC=C3)C=C12
This item is not returnable. View return policy
145915-58-8
C20H15N3O2
329.36
MFCD00236442
AAALVYBICLMAMA-UHFFFAOYSA-N
daph, 4,5-dianilinophthalimide, 5,6-bis phenylamino-1h-isoindole-1,3 2h-dione, 5,6-dianilino-1h-isoindole-1,3 2h-dione, unii-z13d008fz2, d03qtp, daph-1, 4,5-bis anilino phthalimide, 4,5-bis-anilino phthalimide, 1h-isoindole-1,3 2h-dione, 5,6-bis phenylamino
1697
CHEBI:53110
5,6-bis(phenylamino)-2,3-dihydro-1H-isoindole-1,3-dione
O=C1NC(=O)C2=CC(NC3=CC=CC=C3)=C(NC3=CC=CC=C3)C=C12

EGFR inhibitor. Also inhibits Aβ42 fibril formation

Selective inhibitor of the epidermal growth factor receptor (EGFR) (IC50 = 0.3 μM in vitro). Also inhibits and reverses the formation of Aβ42 fibers associated with Alzheimer's disease. Eliminates calcium influx potential when coincubated with amyloid β-peptide (1-42) (Cat.No. 1428); reduces neurotoxicity by blocking Ca2+ influx into neuronal cells.
Quantity 10 mg
Formula Weight 329.35
Percent Purity >99%
Chemical Name or Material CGP 52411
Recommended Storage Store at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit