Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CGP 55845 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent, selective GABAB receptor antagonist (IC50 = 5 nM) that prevents agonist binding (pKi = 8.35) and inhibits GABA and glutamate release (pEC50 values are 8.08 and 7.85 respectively). Inhibits GABAB responses to baclofen (IC50 = 130 nM in an isoproterenol assay) and potentiates the hypoglycemic response to glucose in vitro.
Specifications
Quantity | 10 mg |
Formula Weight | 438.71 |
Percent Purity | >98% |
Chemical Name or Material | CGP 55845 hydrochloride |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction