Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CGREF1 Rabbit anti-Human, DyLight 680, Clone: 1, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP290214FR

Catalog No. NB038160


Add to cart

Description

Description

CGREF1 Monoclonal antibody specifically detects CGREF1 in Human samples. It is validated for Western Blot, ELISA
Specifications

Specifications

CGREF1
1
Western Blot, ELISA
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Western Blot
DyLight 680
50mM Sodium Borate
cell growth regulator with EF hand domain protein 1, cell growth regulator with EF-hand domain 1, Cell growth regulatory gene 11 protein, CGR11cell growth regulator with EF hand domain 1, hydrophobestin
Recombinant Human CGREF1 Protein
Protein A purified
RUO
Primary
10669
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit