Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CGRP Chicken anti-Rat, Polyclonal, Neuromics™ Non-distribution product as customer accommodation.

Chicken Polyclonal Antibody

$277.76 - $602.68

Specifications

Antigen CGRP
Concentration 1 mg/mL
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-209-9989 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
CH14100150UL
150 μL
Each for $602.68
Add to cart
 
50-209-9990 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
CH1410050UL
50 μL
Each for $277.76
Add to cart
 
Description

Description

CGRP Polyclonal antibody specifically detects CGRP in Rat samples. It is validated for Immunohistochemistry
Specifications

Specifications

CGRP
Immunohistochemistry
Unconjugated
Chicken
Rat
P01256, P01257
24241
IgY
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Calca
1 mg/mL
Polyclonal
Liquid, Purified
RUO
PBS with 0.05% sodium azide
Alpha-type CGRP; CA; Cal1; CAL6; Calc; CALC1; Calca; Calcitonin; calcitonin 1; Calcitonin carboxyl-terminal peptide; Calcitonin gene-related peptide 1; Calcitonin gene-related peptide I; calcitonin related polypeptide alpha; calcitonin/calcitonin-related polypeptide, alpha; calcitonin; calcitonin gene-related peptide 1; calcitonin-related polypeptide alpha; CCALCI; CCP; Cgrp; CGRP1; CGRP-1; CGRP-I; Ct; Ctn; Katacalcin; KC; MGC126648; PCT; PDN-21; prepro-alpha-calcitonin gene-related peptide; RAT calcitonin; RATCAL6
KDNFVPTNVGSEAF-NH2
Polyclonal
Affinity chromatography
Antibody
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit