Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Procalcitonin Mouse anti-Human, Rat, Janelia Fluor 549, Clone: 5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP105164JF549

Catalog No. NBP15164549


Add to cart

Description

Description

Procalcitonin Monoclonal antibody specifically detects Procalcitonin in Human, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications

Specifications

Procalcitonin
5
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Purified
CALCA
Synthetic human alfa-CGRP coupled to carrier
Protein A or G purified
Store at 4°C in the dark.
Monoclonal
Based on inhibition ELISA, ABS 026-05 shows full cross-reactivity with rat alpha-CGRP and about about 50% cross-reactivity with human beta-CGRP.
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
CALC1, CALCA, Calcitonin 1, calcitonin gene-related peptide 1, CGRP, Katacalcin
Mouse
IgG1 Kappa
0.1mL
Primary
796
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit