Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CGS 20625, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
111205-55-1
C18H19N3O2
309.369
UBLXQFIFWUEVGJ-UHFFFAOYSA-N
unii-k9q4r9s81o,5,6,7,8,9,10-hexahydro-2-4-methoxyphenyl cyclohepta b pyrazolo 3,4-d pyridin-3 2h-one,cyclohepta b pyrazolo 3,4-d pyridin-3 2h-one,5,6,7,8,9,10-hexahydro-2-4-methoxyphenyl,acmc-20cu5p,2-4-methoxyphenyl-1,6,7,8,9,10-hexahydrocyclohepta b pyrazolo 3,4-d pyridin-3 2h-one,2-4-methoxyphenyl-5,6,7,8,9,10-hexahydrocyclohepta b pyrazolo 3,4-d pyridin-3 2h-one,4-4-methoxyphenyl-3,4,8-triazatricyclo 7.5.0.0 2 ,? tetradeca-1,6,8-trien-5-one,cyclohepta b pyrazolo 3,4-d pyridin-3 2h-one, 5,6,7,8,9,10-hexahydro-2-4-methoxyphenyl
163844
COC1=CC=C(C=C1)N2C(=O)C3=CN=C4CCCCCC4=C3N2
This item is not returnable. View return policy
111205-55-1
C18H19N3O2
309.369
UBLXQFIFWUEVGJ-UHFFFAOYSA-N
unii-k9q4r9s81o,5,6,7,8,9,10-hexahydro-2-4-methoxyphenyl cyclohepta b pyrazolo 3,4-d pyridin-3 2h-one,cyclohepta b pyrazolo 3,4-d pyridin-3 2h-one,5,6,7,8,9,10-hexahydro-2-4-methoxyphenyl,acmc-20cu5p,2-4-methoxyphenyl-1,6,7,8,9,10-hexahydrocyclohepta b pyrazolo 3,4-d pyridin-3 2h-one,2-4-methoxyphenyl-5,6,7,8,9,10-hexahydrocyclohepta b pyrazolo 3,4-d pyridin-3 2h-one,4-4-methoxyphenyl-3,4,8-triazatricyclo 7.5.0.0 2 ,? tetradeca-1,6,8-trien-5-one,cyclohepta b pyrazolo 3,4-d pyridin-3 2h-one, 5,6,7,8,9,10-hexahydro-2-4-methoxyphenyl
163844
COC1=CC=C(C=C1)N2C(=O)C3=CN=C4CCCCCC4=C3N2

Selective central benzodiazepine receptor partial agonist

Selective, partial agonist for the benzodiazepine binding site of the GABA A receptor. Potently inhibits [3H]-flunitrazepam binding to central benzodiazepine receptors (IC 50 = 1.3 nM) and displays weak affinity for peripheral benzodiazepine (IC 50 = 0.68 - 2.25 μM) and GABA binding sites (IC 50 > 10000 μM). Displays anxiolytic activity in vivo following oral administration. .
Quantity 50mg
Formula Weight 309.36
Percent Purity >99%
Chemical Name or Material CGS 20625

Recommended Storage : Store at +4°C