Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CGS 20625, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective, partial agonist for the benzodiazepine binding site of the GABAA receptor. Potently inhibits [3H]-flunitrazepam binding to central benzodiazepine receptors (IC50 = 1.3 nM) and displays weak affinity for peripheral benzodiazepine (IC50 = 0.68 - 2.25 μM) and GABA binding sites (IC50 > 10000 μM). Displays anxiolytic activity in vivo following oral administration. .
Specifications
Quantity | 10 mg |
Synonym | unii-k9q4r9s81o, 5,6,7,8,9,10-hexahydro-2-4-methoxyphenyl cyclohepta b pyrazolo 3,4-d pyridin-3 2h-one, cyclohepta b pyrazolo 3,4-d pyridin-3 2h-one,5,6,7,8,9,10-hexahydro-2-4-methoxyphenyl, acmc-20cu5p, 2-4-methoxyphenyl-1,6,7,8,9,10-hexahydrocyclohepta b pyrazolo 3,4-d pyridin-3 2h-one, 2-4-methoxyphenyl-5,6,7,8,9,10-hexahydrocyclohepta b pyrazolo 3,4-d pyridin-3 2h-one, 4-4-methoxyphenyl-3,4,8-triazatricyclo 7.5.0.0 2 ,? tetradeca-1,6,8-trien-5-one, cyclohepta b pyrazolo 3,4-d pyridin-3 2h-one, 5,6,7,8,9,10-hexahydro-2-4-methoxyphenyl |
Formula Weight | 309.36 |
Percent Purity | >99% |
Chemical Name or Material | CGS 20625 |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction