Learn More
Azzota Corp CH-5183284 (Debio-1347) DFS Item
Manufacturer:  Azzota Corp C1108
CH-5183284 (Debio-1347) is an orally bioavailable inhibitor of the fibroblast growth factor receptor subtypes 1 (FGFR-1), 2 (FGFR-2) and 3 (FGFR-3), with potential antineoplastic activity. Please request if need more quantities
Catalog No. 50-161-3091