Learn More
ChABC Mouse anti-Non-species specific, DyLight 350, Clone: 10000000000, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP196141UV
Catalog No. NBP196141UV
Add to cart
Description
Description
ChABC Monoclonal antibody specifically detects ChABC in Rat, Non-species specific samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
ChABC
10000000000
Western Blot, ELISA
Purified
Mouse
IgG
0.1 ml
Primary
Non-species specific
Western Blot, ELISA
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
Chondroitinase ABC
Chondroitinase ABC
Protein G purified
Store at 4C in the dark.
Monoclonal