Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHADL Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548781

Catalog No. PA548781

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Potential negative modulator of chondrocyte differentiation. Inhibits collagen fibrillogenesis in vitro. May influence chondrocyte ft.s differentiation by acting on its cellular collagenous microenvironment.
Specifications

Specifications

CHADL
Polyclonal
PBS with 0.09% sodium azide
Q6NUI6
Chondroadherin-like protein, CHADL, SLRR4B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
150356
Western Blot
Unconjugated
CHADL
Liquid
CHADL
KLH conjugated synthetic peptide between 406-434 amino acids from the Central region of human CHADL.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit