Chalcopyrite, Alfa Aesar™

Manufacturer: Alfa Aesar 4253303

Quantity 1g

 View more versions of this product

Catalog No. AA4253303

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1308-56-1
    MDL Number MFCD00064817
    Chemical Name or Material Chalcopyrite

    Specifications

    Quantity 1g
    MDL Number MFCD00064817
    CAS 1308-56-1
    Chemical Name or Material Chalcopyrite

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit