Learn More
Eisco™ Chalcopyrite Mineral Specimen
SCI_ED
Catalog No. s27573 Shop All Eisco Products
Click to view available options
Quantity:
1/Ea.
12/Pk.
From the same family as fool's gold, Chalcopyrite is a yellow crystalline mineral consisting of a sulfide of copper and iron. It's the principal ore of copper formed in hydrothermal veins.
  • Correlates well to Next Generation Science Standards Evidence
  • Chalcopyrite's formation exhibits fracture and has a brassy luster which make it an excellent tool for studying economic geology
  • Great for us in comparative identification exercises
Specifications
Product Type Mineral
Grade K to 16
Quantity 1/Ea.
Specimen Size 2 to 3 cm
Weight (Metric) 0.2 kg
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.