Learn More
CHAMP1 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304216A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Required for proper alignment of chromosomes at metaphase and their accurate segregation during mitosis. Involved in the maintece of spindle microtubules attachment to the kinetochore during sister chromatid biorientation. May recruit CENPE and CENPF to the kinetochore.Specifications
CHAMP1 | |
Polyclonal | |
Unconjugated | |
CHAMP1 | |
C13orf8, CAMP, CHAMP, chromosome 13 open reading frame 8, chromosome alignment-maintaining phosphoprotein, MRD40, zinc finger protein 828, ZNF828 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
101994, 283489 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q8K327, Q96JM3 | |
CHAMP1 | |
Between 1 and 50 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.