Learn More
ChannelLock™ Adjustable Wrenches
Perfectly parallel jaw surfaces
Manufacturer:  Nasco 140806WCLAM
 View more versions of this product
Catalog No. 18-861-276
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
6 x 1.89 in. (15.24 x 4.8cm)
15°
6 in.
Chrome Vanadium Steel
0.11kg
4.8cm
I-Beam
0.93 in.
15.24cm
0.26 lb.
1.89 in.
Jaw Opening Adjustable wrench