Learn More
ORS Nasco ChannelLock™ Lineman's Pliers
Perfectly mated cutter, heat-treated to hold its edge
Manufacturer:  ORS Nasco 140347BULK
 View more versions of this product
Catalog No. 18-861-268
Add to cart
Description
Description
  • Used for both insulated and non-insulated terminals
  • Durable and smooth operating box joint construction
  • A useful crushing area between handles and joint
  • Electrocoated protective finish to prevent rusting
  • Cross-hatched jaw tip for maximum gripping
Specifications
Specifications
7 in.
High Carbon Steel
Round Nose
1.28 in.
0.62 lb.
17.78 cm
ChannelLock™ blue comfort
3.25 cm
Cross-hatched, Rounded Nose