Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHAPS, GE Healthcare Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
1g
75621-03-3
C32H58N2O7S
614.883
UMCMPZBLKLEWAF-GBSSQDIOSA-N
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate
134129639
3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate
CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C
75621-03-3
C32H58N2O7S
614.883
UMCMPZBLKLEWAF-GBSSQDIOSA-N
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate
134129639
3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate
CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C

Use CHAPS and Triton X-100 as detergents in IEF. CHAPS, GE Healthcare includes a nonionic or zwitterionic detergent in the sample solution to ensure complete sample solubilization and to prevent aggregation through hydrophobic interactions.

  • Used for IEF, PAGE
  • Nonionic or zwitterionic detergent
  • Analytic specification of raw material supplied with the product
  • Partially soluble in cold and hot water 

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Formula Weight 614.88
Quantity 1g
Chemical Name or Material CHAPS