CHAPS, Ultrapure Bioreagent, J.T.Baker™

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

Manufacturer: J.T.Baker 414500

Packaging POLYSTORMOR
Quantity 5g

 View more versions of this product

Catalog No. 02-002-890

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 75621-03-3
    Molecular Formula C32H58N2O7S
    Formula Weight 614.89
    Synonym 3-(3-cholamidopropyl)dimethylammonio-1-propanesulfonate
    Chemical Name or Material CHAPS
    Grade Ultrapure Bioreagent

    Specifications

    Formula Weight 614.89
    CAS 75621-03-3
    Packaging POLYSTORMOR
    Molecular Formula C32H58N2O7S
    Synonym 3-(3-cholamidopropyl)dimethylammonio-1-propanesulfonate
    Quantity 5g
    Chemical Name or Material CHAPS
    Grade Ultrapure Bioreagent

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit