Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Characterized Fetal Bovine Serum, Canadian Origin

GE Healthcare™ Characterized Fetal Bovine Serum, Canadian Origin enhances the growth of cells with Canadian sourced HyClone Characterized Fetal Bovine Serum.

Manufacturer:  GE Healthcare SH30396.03IR

Catalog No. SH3039603IR


Add to cart

Description

Description

  • Characterized Fetal Bovine Serum (FBS), Canadian origin, is filtered using 100 nm (0.1 µm) pore size-rated filters and processed using true pool technology to ensure consistency in the quality of the serum (Fig 1). Characterized FBS is widely used by professionals requiring an extensive biochemical profile and full traceability back to the original source. Characterized FBS, Canadian Origin is compliant with current good manufacturing practices (cGMP).
  • Characterized FBS is filtered through three sequential 100 nm (0.1 µm) pore size-rated filters. Before dispensing, each lot of serum is pooled to ensure uniformity and consistency between bottles (true pool technology). Characterized FBS is assayed for gamma globulin, alkaline phosphatase, lactate dehydrogenase, glutamic pyruvic transaminase (SBPT), glutamic oxaloacetic transaminase (SGOT), pH, total protein, albumin, blood urea nitrogen, creatinine, total bilirubin, sodium, potassium, calcium, chloride, inorganic phosphorus, osmolality, iron, total iron binding capacity (TIBC), percent iron saturation, glucose, and IgG. Included assays are subject to change without notice.
Specifications

Specifications

Yes
≤ 25 EU/ml
≤ 25 mg/dl
No
Filtered though sequential 100 nm pore-size rated filters; Irradiated.
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
Bluetongue Bovine Adenovirus Bovine Parvovirus Bovine RSV BVDV Cytopathogenic Agents Hemadsorbing Agents Rabies Virus Reovirus 
Gamma Globulin Alkaline Phosphatase Lactate Dehydrogenase SGPT SGOT pH Albumin Blood Urea Nitrogen Creatinine Total Bilirubin Sodium Potassium Calcium Chloride Inorganic Phosphorus Iron TIBC Percent Saturation Glucose IgG 
Characterized
Canada
Irradiated
500 ml
Yes
Documents
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit