Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Charcoal, Activated, Norit(R) RBHG, 3 mm Pellet, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Amber Glass Bottle
Poly Pail
7440-44-0
7440-44-0
Packaging Amber Glass Bottle
Quantity 500 g
CAS Max % 1
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.