Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Charcoal, Activated, Powder, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
40 lb.
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
7440-44-0
7440-44-0
Packaging Fiber Drum
Quantity 40 lb.
CAS Max % 1
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.