Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Chargeswitch™ Pro Filter Plasmid Maxi Kit

Chargeswitch™ Pro Filter Plasmid Maxi Kit

$445.00 - $800.00

Specifications

For Use With (Application) Next-Generation Sequencing, Transfection, Cloning, Sequencing, Transformation, Nucleic Acid Labeling, PCR, In Vitro Transcription
Isolation Technology ChargeSwitch™ Chemistry
Purity Transfection Grade
Yield 800 µg
Format Spin Column
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
CS31106
Encompass
View Documents
Invitrogen™
CS31106
10 Columns
Each for $445.00
Add to cart
 
CS31107
Encompass
View Documents
Invitrogen™
CS31107
25 Columns
Each for $800.00
Add to cart
 
Description

Description

The Chargeswitch™ -Pro Filter Plasmid Maxi Kit provides high yield and high purity plasmid with the fastest and the simplest protocol.
The ChargeSwitch™-Pro Plasmid Filter Mini, Midi, Maxi Kits generates high-quality plasmid DNA without the use of harsh or potentially interfering reagents, for better success in downstream applications. The system is based upon a novel dual⁄nested column design which facilitate the fastest and simplest protocol for high quality plasmid. The system is composed of two columns. The inner flat bottomed column, also referred to as “lysate clarification” column, contains filter materials suitable for rapid clarification of bacterial lysate. The outer luer bottomed column, also referred to as “binding” column, contains ChargeSwitch™ (CST) derivatized membrane that immediately binds plasmid DNA from the clarified lysate. This nested-column design enables simultaneous clarification and DNA binding in a single short centrifugation (or vacuum) step. The combined advantages of novel, optimized plastics and a unique chemistry are an extreme competitive advantage of speed and convenience.



To accomplish this, the ChargeSwitch™-Pro Filter Kit provides:

  • 100% ethanol-free, guanidinium-free, organics-free, aqueous protocol for a safer, more streamlined method
  • Cleaner purified product for improved performance in downstream applications
  • A novel Dual⁄nested column design for simultaneous lysate clearing and binding step which offers the fastest and the simplest protocol
  • Endotoxin units < 10 EU ⁄ ug DNA for
  • Vacuum Elution can be done with the EveryPrep™ Universal Vacuum Manifold (SKU # K211101)



    How it works
    The ChargeSwitch™ technology replaces the use of ethanol, chaotropic salts, or organic solvents for nucleic acid purification with a simple ion-exchange mechanism. At low pH values (<6.5) the ChargeSwitch™ surface ligand is positively charged and binds plasmid DNA. Raising the pH (>8.5) neutralizes the charge and elutes the bound nucleic acid. The ChargeSwitch™ protocol retains the ease and familiarity of the common silica spin column protocol with a binding, washing, and elution step, making adapting to the ChargeSwitch™ method effortless.
  • Specifications

    Specifications

    Next-Generation Sequencing, Transfection, Cloning, Sequencing, Transformation, Nucleic Acid Labeling, PCR, In Vitro Transcription
    Transfection Grade
    Spin Column
    ChargeSwitch™ Chemistry
    800 µg
    Safety and Handling

    Safety and Handling

    SDS
    Product Certifications

    For Research Use Only. Not for use in diagnostic procedures.

    Provide Content Correction

    The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit